Search
Our qIP Protein Interaction Kits use anti-HA or anti-Myc beads (agarose or magnetic) and a sensitive luciferase assay system to co-immunoprecipitate (co-IP) and quantify interactions between epitope-tagged and TurboLuc luciferase enzyme (Tluc) tagged protein pairs expressed in mammalian cells. The quantitative immunoprecipitation (qIP) system uses Tluc enzyme to accurately and precisely reflect the abundance of a specific co-IP product, to help avoid the need for time-consuming gel electrophoresis, western blotting, and band densitometry.
Learn more about
An agarose bead format of our qIP kit, using HA tagging
Exploit the convenience of magnetic bead-mediated qIP
Recover and quantify proteins that bind to your Myc-tagged bait protein
Magnetic beads make quantitative immunoprecipitation fast and robust
For Research Use Only. Not for use in diagnostic procedures.