• More thermostable than AMV RT and M-MLV RT
  • Excellent target to evaluate antiviral agents
  • Highly sensitive enzyme for RT-PCR applications
  • Ideal for reverse transcribing transcripts with high G/C content
Like M-MLV reverse transcriptase (RT) and AMV RT, Ambion's HIV RT synthesizes cDNA from single-stranded RNA template in the presence of primer, Mg 2+, and deoxyribonucleotides. HIV-RT is a recombinant enzyme purified to homogeneity from E. coli. HIV RT is a heterodimeric protein which is comprised of a 66 kDa and a 51 kDa subunit.
HIV RT is More Thermostable than M-MLV RT and AMV RT
HIV RT performs as well as M-MLV RT and AMV RT at 37°C and 42°C. However, the enzyme retains a significantly higher fraction of its activity at 50°C than the other RTs. As shown in Figure 1, HIV RT converts a much larger proportion of a synthetic RNA transcript to cDNA than M-MLV RT and AMV RT even after the reaction has progressed for 1 hour. Thus, when working with RNA transcripts that are refractory to efficient cDNA synthesis or when the transcript being studied has a high GC content, HIV RT is a superior alternative to M-MLV RT and AMV RT.
Ambion's HIV RT is a high quality enzyme that has been tested for contaminating endonuclease and exonuclease activity. The enzyme is available in a convenient 500 U size. A 10X Reaction Buffer is supplied with the enzyme.
Figure 1. HIV RT Outperforms MMLV and AMV RT Enzymes in cDNA Synthesis at 50ºC. A synthetic RNA transcript for Xef-1 (1 µg) was reverse transcribed with oligo(dT) primers in a 20 µl reaction using 5 µg/µl M-MLV RT (Ambion), 0.5 U/µl AMV RT (Seikagaku), or 0.5 µg/µl HIV RT (Ambion) for the indicated times. Reactions were performed using the manufacturer-supplied reaction buffer in the presence of *32-P dUTP and 0.5 U/µl ribonuclease inhibitor. The percentage cDNA conversion was determined by measuring the amount of cDNA made via scintillation counting.
Learn more about reverse transcription or use our selection tool to identify the best reverse transcriptase for your needs.
References
  1. Sorbo BH (1953) Acta Chem Scand 7: 1129.
  2. Kudlicki W, Odom OW, Kramer G, Hardesty B (1994) Activation and release of enzymatic ally inactive, full-length Rhodanese that is bound to ribosomes as peptide-tRNA. J Biol Chem 269(24): 16549-165