Search
Thermo Scientific SamStat-30 Representative Slurry Sampling Stations and AnStat-330 Online Sampling and Elemental Analysis Stations both provide periodic composite metallurgical samples for accounting while providing online elemental analysis. Which model best fits your needs? Use this comparison chart to see which system is right for you.
Still have questions? We're here to help!
| Slurry Samplers | |||||||
| Product Family | Differentiating Features | Metallurgical Composite Sampler | Analyzer Type | Pebble Screen | High Froth | Sample Take-off | Distributor |
| SamStat-30 | Upgradable to AnStat | Y | N | N | N | N | N |
| SamStat-30F | Upgradable to AnStat | Y | N | N | Y | N | N |
| SamStat-30D | Upgradable to AnStat | Y | N | N | N | Y | |
| SamStat-30T | Upgradable to AnStat | Y | N | N | N | Y | N |
| Sam-Stat30P | Upgradable to AnStat | Y | N | Y | N | N | N |
| SamStat-30DP | Upgradable to AnStat | Y | N | Y | N | N | Y |
| SamStat-30DT | Upgradable to AnStat | Y | N | N | N | Y | Y |
| SamStat-30PT | Upgradable to AnStat | Y | N | Y | N | Y | N |
| SamStat-30DPT | Upgradable to AnStat | Y | N | Y | N | Y | Y |
| SamStat-30C | Continuous Flow + Incremental Sampling | N | N | N | N | N | N |
| SamStat-30CP | Continuous Flow + Pebble Screen | N | N | Y | N | N | N |
| SamStat-30CF | Continuous Flow + Froth | N | N | N | Y | N | N |
| SamStat-30CT | Continuous Flow + Take off point | N | N | N | N | Y | N |
| SamStat-30CD | Continuous Flow + distributor | N | N | N | N | N | Y |
| SamStat-30CDP | Continuous Flow + distributor + Pebble Screen | N | N | Y | N | N | Y |
| SamStat-30CDT | Continuous Flow + distributor + take off point | N | N | N | N | Y | Y |
| SamStat-30CPT | Continuous Flow + Pebble Screen + Take off point | N | N | Y | N | Y | N |
| SamStat-30CDPT | Continuous Flow + distributor + Pebble Screen + Take off | N | N | Y | N | Y | Y |
| SamStat-30R | Primary Sampler for Particle Size Monitor + Reverse sampling | N | N | N | N | N | N |
| GF-Samp | Gravity Pipe Sampler | N | N | N | N | N | N |
| PP-Samp | Pressure Pipe Sampler | N | N | N | N | N | N |
| X-Cut Sampler | Cross Cut Composite Sampler | Y | N | N | N | N | N |
| Dedicated XRF Analyzer Stations | |||||||
| Product Family | Differentiating Features | Metallurgical Composite Sampler | Analyzer Type | Pebble Screen | High Froth | Sample Take-off | Distributor |
| AnStat-330 | Generation 3 Liquid Nitrogen Free | Y | XRF | N | N | N | N |
| AnStat-330P | Generation 3 Liquid Nitrogen Free + Pebble Screen | Y | XRF | Y | N | N | N |
| AnStat-330F | Generation 3 Liquid Nitrogen Free + Froth stream | Y | XRF | N | Y | N | N |
| AnStat-330T | Generation 3 Liquid Nitrogen Free + Take off point | Y | XRF | N | N | Y | N |
| AnStat-330T2 | Generation 3 Liquid Nitrogen Free + Twin Inlet | Y | XRF | N | N | Y-2 | N |
| AnStat-330T3 | Generation 3 Liquid Nitrogen Free + Triple Inlet | Y | XRF | N | N | Y-3 | N |
| AnStat-330D | Generation 3 Liquid Nitrogen Free + distributor | Y | XRF | N | N | N | Y |
| AnStat-330PT | Generation 3 Liquid Nitrogen Free + Pebble screen + Take off point | Y | XRF | Y | N | Y | N |
| AnStat-330DP | Generation 3 Liquid Nitrogen Free + Distributor + Pebble screen | Y | XRF | Y | N | N | Y |
| AnStat-330DT | Generation 3 Liquid Nitrogen Free + Distributor + Take off point | Y | XRF | N | N | Y | Y |
| AnStat-330DPT | Generation 3 Liquid Nitrogen Free + distributor + Pebble screen + Take off point | Y | XRF | Y | N | Y | Y |
| On-line Analyzers | |||||||
| Product Family | Differentiating Features | Process Type | Analysis Type | Method | Source | Streams | Composite & Calibration Sampler |
| AnStat-330 | Sampler/Analyzer | Slurry | Elemental | XRF | Isotope | 1 | Y |
| MSA-330 | Multi-stream Centralized | Slurry | Elemental | XRF | Isotope | 3-12 | Y |
| GS-Omni | Multi-stream Centralized | Slurry | Elemental | PGNAA | Isotope | 2-8 | Y |
| PSM-500 | Sample Conditioner/Analyzer | Slurry | Particle Size | Ultrasonic Absorption | n/a | 1-3 | Y |
| CB-Omni Agile | Single stream | Conveyer Belt | Elemental | PGNAA | Isotope nGen | 1 | Y |
| Slurry Samplers | |||||||
| Product Family | Differentiating Features | Metallurgical Composite Sampler | Analyzer Type | Pebble Screen | High Froth | Sample Take-off | Distributor |
| SamStat-30 | Upgradable to AnStat | Y | N | N | N | N | N |
| SamStat-30F | Upgradable to AnStat | Y | N | N | Y | N | N |
| SamStat-30D | Upgradable to AnStat | Y | N | N | N | Y | |
| SamStat-30T | Upgradable to AnStat | Y | N | N | N | Y | N |
| Sam-Stat30P | Upgradable to AnStat | Y | N | Y | N | N | N |
| SamStat-30DP | Upgradable to AnStat | Y | N | Y | N | N | Y |
| SamStat-30DT | Upgradable to AnStat | Y | N | N | N | Y | Y |
| SamStat-30PT | Upgradable to AnStat | Y | N | Y | N | Y | N |
| SamStat-30DPT | Upgradable to AnStat | Y | N | Y | N | Y | Y |
| SamStat-30C | Continuous Flow + Incremental Sampling | N | N | N | N | N | N |
| SamStat-30CP | Continuous Flow + Pebble Screen | N | N | Y | N | N | N |
| SamStat-30CF | Continuous Flow + Froth | N | N | N | Y | N | N |
| SamStat-30CT | Continuous Flow + Take off point | N | N | N | N | Y | N |
| SamStat-30CD | Continuous Flow + distributor | N | N | N | N | N | Y |
| SamStat-30CDP | Continuous Flow + distributor + Pebble Screen | N | N | Y | N | N | Y |
| SamStat-30CDT | Continuous Flow + distributor + take off point | N | N | N | N | Y | Y |
| SamStat-30CPT | Continuous Flow + Pebble Screen + Take off point | N | N | Y | N | Y | N |
| SamStat-30CDPT | Continuous Flow + distributor + Pebble Screen + Take off | N | N | Y | N | Y | Y |
| SamStat-30R | Primary Sampler for Particle Size Monitor + Reverse sampling | N | N | N | N | N | N |
| GF-Samp | Gravity Pipe Sampler | N | N | N | N | N | N |
| PP-Samp | Pressure Pipe Sampler | N | N | N | N | N | N |
| X-Cut Sampler | Cross Cut Composite Sampler | Y | N | N | N | N | N |
| Dedicated XRF Analyzer Stations | |||||||
| Product Family | Differentiating Features | Metallurgical Composite Sampler | Analyzer Type | Pebble Screen | High Froth | Sample Take-off | Distributor |
| AnStat-330 | Generation 3 Liquid Nitrogen Free | Y | XRF | N | N | N | N |
| AnStat-330P | Generation 3 Liquid Nitrogen Free + Pebble Screen | Y | XRF | Y | N | N | N |
| AnStat-330F | Generation 3 Liquid Nitrogen Free + Froth stream | Y | XRF | N | Y | N | N |
| AnStat-330T | Generation 3 Liquid Nitrogen Free + Take off point | Y | XRF | N | N | Y | N |
| AnStat-330T2 | Generation 3 Liquid Nitrogen Free + Twin Inlet | Y | XRF | N | N | Y-2 | N |
| AnStat-330T3 | Generation 3 Liquid Nitrogen Free + Triple Inlet | Y | XRF | N | N | Y-3 | N |
| AnStat-330D | Generation 3 Liquid Nitrogen Free + distributor | Y | XRF | N | N | N | Y |
| AnStat-330PT | Generation 3 Liquid Nitrogen Free + Pebble screen + Take off point | Y | XRF | Y | N | Y | N |
| AnStat-330DP | Generation 3 Liquid Nitrogen Free + Distributor + Pebble screen | Y | XRF | Y | N | N | Y |
| AnStat-330DT | Generation 3 Liquid Nitrogen Free + Distributor + Take off point | Y | XRF | N | N | Y | Y |
| AnStat-330DPT | Generation 3 Liquid Nitrogen Free + distributor + Pebble screen + Take off point | Y | XRF | Y | N | Y | Y |
| On-line Analyzers | |||||||
| Product Family | Differentiating Features | Process Type | Analysis Type | Method | Source | Streams | Composite & Calibration Sampler |
| AnStat-330 | Sampler/Analyzer | Slurry | Elemental | XRF | Isotope | 1 | Y |
| MSA-330 | Multi-stream Centralized | Slurry | Elemental | XRF | Isotope | 3-12 | Y |
| GS-Omni | Multi-stream Centralized | Slurry | Elemental | PGNAA | Isotope | 2-8 | Y |
| PSM-500 | Sample Conditioner/Analyzer | Slurry | Particle Size | Ultrasonic Absorption | n/a | 1-3 | Y |
| CB-Omni Agile | Single stream | Conveyer Belt | Elemental | PGNAA | Isotope nGen | 1 | Y |