Search
| Components | Molecular Weight | Concentration (mg/L) | mM |
|---|---|---|---|
| Inorganic Salts | |||
| Potassium Chloride (KCl) | 75.0 | 4000.0 | 53.333332 |
| Potassium Phosphate monobasic (KH2PO4) | 136.0 | 600.0 | 4.4117646 |
| Sodium Chloride (NaCl) | 58.0 | 80000.0 | 1379.3103 |
| Sodium Phosphate dibasic (Na2HPO4-7H2O) | 268.0 | 900.0 | 3.358209 |
| Other Components | |||
| D-Glucose (Dextrose) | 180.0 | 10000.0 | 55.555557 |
1. Hanks, J.H. and Wallace, R.E. (1949) Relation of Oxygen and Temperature in the Preservation of Tissues by Refrigeration. Proc. Soc. Exp. Biol. Med., 71:196.
14170 - HBSS, no calcium, no magnesium
Cat No:
14170070
,
14170088
,
14170112
,
14170120
,
14170138
,
14170146
,
14170161
14175 - HBSS, no calcium, no magnesium, no phenol red
Cat No:
14175046
,
14175053
,
14175079
,
14175095
,
14175103
,
14175129
,
14175137
,
14175145