Multidrop Pico Digital Dispensers
Thermo Scientific Multidrop Pico 1 and 8 Surfactant Free Digital Dispensers offer enhanced dispensing precision with volumes between 11 pL and 10 μL in any well, eliminating the need for manual dilution while providing significant cost and waste reductions. Designed to enable assay miniaturization, boost productivity, and improve accuracy of results, these dispensers are ideal for a broad array of low-volume applications such as qPCR, dose response curves, drug screening assays.
Features
  1. Assay Miniaturization: Enables the use of up to 10 times less reagent and sample, reducing costs and conserving valuable materials.
  2. Digital Dispensing: Provides precise, consistent fluid dispensing down to 11 pL, eliminating the need for manual dilution.
  3. Improved Efficiency: Achieves typical dose experiments in just 15 seconds, dramatically faster than traditional methods.
  4. Non-Contact Dispensing: Reduces the risk of cross-contamination and carry-over with disposable dispensing heads.
  5. PicoIT Software: Simplifies experiment design with flexible, intuitive tools and application-focused wizards, ensuring consistent results and full traceability of reagents.
Specifications:
Featured videos
Get a look inside the Pico
Note: This tool is best viewed in Chrome, Firefox or Safari Browsers
Applications
  • qPCR
  • Drug combination studies
  • DMPK
  • Assay development
  • Biochemical assays
  • Secondary screening
  • Cell-based assays
  • Spotting
  • Enzyme profiling
  • Synthetic biology
  • Antibody therapies
  • SAR
PicoIT Software
An easy-to-use Multidrop PicoIT software platform with application-focused wizards and a highlighted loading guide allows effortless and quick plate setup and operation. Available in one- and eight-well formats to support different throughputs and amounts of reagents and samples.
The pre-programmed protocols provide:
  • Pre-selected fluid information
  • Simplified layout including Non-Template Controls 
  • Pre-set reaction volume and plate type
  • Pre-set number of replicates
Ordering Information
Order below or select product to learn more.
For Laboratory Use. It is the customer’s responsibility to ensure that the performance of the product is suitable for customers’ specific uses or applications.