Search
Diomni Design and Analysis (RUO) Software is a feature-rich application solution that provides plate setup, quality checks, results review, and access to a broad selection of real-time PCR (qPCR) application analysis modules in one software application.
Diomni Design and Analysis (RUO) Software offers powerful tools to support your research.
With customizable analysis options, Diomni Design and Analysis (RUO) Software provides the flexibility to adapt to your research needs.
With more than 25 years of qPCR innovation, Applied Biosystems qPCR instruments continue to lead the way in performance, reliability, and world-class support. Embrace the future of scientific discovery with our comprehensive collection of QuantStudio qPCR systems. Our continuous innovation in qPCR technology yields ongoing advancements to support performance across the QuantStudio instrument family, including:
The security, audit, and electronic Signature (SAE) module is designed to assist compliance with 21 CFR Part 11 guidelines on electronic records security and to prevent unauthorized access to the instrument. The SAE features are only available with the on-premises deployment solution.
From gene expression to Cq analysis and copy number variation determination, we have developed robust analysis software for nearly every real-time PCR application. Plus, all our analysis tools have been optimized for use with our TaqMan Assays on Applied Biosystems Real-time PCR instruments.
| Instrument compatibility for analysis | QuantStudio 1 / 3 / 5 QuantStudio 6 / 7 Flex QuantStudio 6 / 7 Pro QuantStudio 12K Flex 7500 / 7500 F 7900 HT StepOne / StepOnePlus ViAA 7 |
| Instrument compatibility for plate setup | QuantStudio 1 / 3 / 5 QuantStudio 6 / 7 Flex QauntStudio 6 / 7 Pro QuantStudio 12K Flex |
| Instrument control compatibility | QuantStudio 6 / 7 Pro AmpQuest 96 |
| Block types | 96-well 0.1mL 96-well 0.2mL 384-well OpenArray TaqMan Array Card |
| Run modes | Fast Standard |
| Analysis modules | Standard curve (single plate analysis) Genotyping (single and multiplate analysis) Presence absence (single plate analysis) Relative quantification (single plate analysis) |
| Server operating system | Microsoft Windows™ 10 or Microsoft Windows 11 |
| Client operating system | Users may access server from any Windows OS or Mac OS |
| Supported browsers | Google Chrome™ Microsoft Edge™ Apple Safari™ (OS X) Mozilla Firefox™ |
| Security and compliance | SAE administrator console (on-premises deployment only) |
| Regulatory environment | RUO (Research Use Only) |
| Desktop | Cloud | On-premises | |
| Description | Download to your PC | Use directly from your browser | Centralized on-premises deployment with multiple user access |
| OS support | Windows 11, Windows 10, Mac OS | Any Windows or Mac OS | Installation: Windows 11, Windows 10 User access: Any Windows or Mac OS |
| Supported browsers | NA | Google Chrome Mozilla Firefox Microsoft Edge |
Google Chrome Mozilla Firefox Microsoft Edge Apple Safari |
| SAE | NA | NA | Security always enabled Audit and eSignature optional |
| Data management | USB or LAN | DataStorage on ThermoFisherConnect | Data Sharing via File Manager within software |
| Software updates | User updates every PC with the software | No customer updating required - latest software available for use | User updates software in one place; centralized installation |