Search
Analyzing aerospace materials and components requires techniques capable of resolving chemical, structural, and surface-level features across multiple length scales. Thermo Fisher Scientific provides a full suite of instruments to support these techniques, including electron microscopes for high-resolution nanoscale imaging, X-ray photoelectron spectroscopy systems for elemental analysis, and focused ion beam scanning electron microscopes that can prepare and image samples all in one system.
SEMs support high-resolution imaging, elemental analysis, and microstructural characterization of aerospace materials. They are widely used for failure analysis, inclusion detection, coating evaluation, and process monitoring across manufacturing and MRO environments.
Thermo Scientific ChemiSEM Systems combine SEM imaging with real-time elemental analysis. These platforms are well suited to automated phase mapping, inclusion analysis, corrosion studies, and rapid quality control in aerospace production environments.
Techniques
Aerospace and defense use cases
Thermo Scientific Phenom ParticleX Desktop SEMs are optimized for particle analysis, supporting automated detection and classification of debris and inclusions. These tools are commonly used for cleanliness analysis, wear debris monitoring, and routine quality assurance.
Techniques
Aerospace and defense use cases
XPS systems deliver surface-sensitive chemical analysis of aerospace materials. They are used to investigate corrosion, oxidation, coatings, passivation layers, surface contamination, and multilayer systems through depth profiling and chemical state analysis.
Techniques
Aerospace and defense use cases
TEMs enable nanoscale and atomic-level analysis of aerospace materials. They are particularly useful for studying precipitates, phase transformations, dislocations, and interfaces in advanced alloys, composites, and additively manufactured parts.
Techniques
Aerospace and defense use cases
FIB-SEMs combine focused ion beam milling and SEM imaging for site-specific sample preparation and 3D analysis. They support cross-sectioning, volumetric reconstruction, and failure analysis of complex aerospace components and coatings.
Techniques
Aerospace and defense use cases