Search
| Components | Molecular Weight | Concentration (mg/L) | mM |
|---|---|---|---|
| Amino Acids | |||
| Glycine | 75.0 | 7.5 | 0.1 |
| L-Alanine | 89.0 | 13.9 | 0.15617977 |
| L-Arginine hydrochloride | 211.0 | 42.1 | 0.19952606 |
| L-Asparagine | 132.0 | 45.0 | 0.3409091 |
| L-Aspartic acid | 133.0 | 19.97 | 0.15015037 |
| L-Cysteine | 121.0 | 31.5 | 0.2603306 |
| L-Glutamic Acid | 147.0 | 22.1 | 0.15034014 |
| L-Glutamine | 146.0 | 219.2 | 1.5013698 |
| L-Histidine hydrochloride-H2O | 210.0 | 20.96 | 0.09980952 |
| L-Hydroxyproline | 131.0 | 19.7 | 0.15038168 |
| L-Isoleucine | 131.0 | 39.36 | 0.300458 |
| L-Leucine | 131.0 | 39.36 | 0.300458 |
| L-Lysine hydrochloride | 183.0 | 36.5 | 0.19945355 |
| L-Methionine | 149.0 | 14.9 | 0.099999994 |
| L-Phenylalanine | 165.0 | 16.5 | 0.1 |
| L-Proline | 115.0 | 17.3 | 0.15043478 |
| L-Serine | 105.0 | 26.3 | 0.25047618 |
| L-Threonine | 119.0 | 17.9 | 0.15042016 |
| L-Tryptophan | 204.0 | 3.1 | 0.0151960775 |
| L-Tyrosine | 181.0 | 18.1 | 0.1 |
| L-Valine | 117.0 | 17.6 | 0.15042736 |
| Vitamins | |||
| Ascorbic Acid | 176.0 | 0.5 | 0.0028409092 |
| Biotin | 244.0 | 0.2 | 8.1967213E-4 |
| Choline chloride | 140.0 | 5.0 | 0.035714287 |
| D-Calcium pantothenate | 477.0 | 0.2 | 4.1928721E-4 |
| Folic Acid | 441.0 | 10.0 | 0.022675738 |
| Niacinamide | 122.0 | 0.5 | 0.0040983604 |
| Nicotinic acid (Niacin) | 123.0 | 0.5 | 0.0040650405 |
| Para-Aminobenzoic Acid | 137.0 | 1.0 | 0.00729927 |
| Pyridoxal hydrochloride | 204.0 | 0.5 | 0.0024509805 |
| Pyridoxine hydrochloride | 206.0 | 0.5 | 0.0024271845 |
| Riboflavin | 376.0 | 0.2 | 5.319149E-4 |
| Thiamine hydrochloride | 337.0 | 0.2 | 5.934718E-4 |
| Vitamin B12 | 1355.0 | 2.0 | 0.0014760147 |
| i-Inositol | 180.0 | 36.0 | 0.2 |
| Inorganic Salts | |||
| Calcium Chloride (CaCl2) (anhyd.) | 111.0 | 100.0 | 0.9009009 |
| Magnesium Sulfate (MgSO4-7H2O) | 246.0 | 200.0 | 0.8130081 |
| Potassium Chloride (KCl) | 75.0 | 400.0 | 5.3333335 |
| Sodium Bicarbonate (NaHCO3) | 84.0 | 2200.0 | 26.190475 |
| Sodium Chloride (NaCl) | 58.0 | 6460.0 | 111.37931 |
| Sodium Phosphate monobasic (NaH2PO4-H2O) | 138.0 | 580.0 | 4.2028985 |
| Other Components | |||
| Bacto-Peptone | 600.0 | Infinity | |
| D-Glucose (Dextrose) | 180.0 | 3000.0 | 16.666666 |
| Glutathione (reduced) | 307.0 | 0.5 | 0.0016286644 |
| Phenol Red | 376.4 | 10.0 | 0.026567481 |
1. Hsu, T.C. and Kellogg, D.S., Jr. (1960) J. Nat. Cancer Inst., 25:221.
2. Iwakata, S. and Grace, J.T., Jr. (1964) N.Y.J. Med., 64:2279.
3. McCoy, T.A., Maxwell, M. and Kruse, P.F. (1959) Proc. Soc. Exper. Biol. Med., 100:115.