Thermo Fisher Scientific at CYTO 2026
June 6–10, 2026
Palm Beach County Convention Center | West Palm Beach, FL, USA
Leading the future of flow cytometry and innovative solutions: please visit us at booth #213
Discover. Connect. Innovate.
Join us at CYTO 2026 to explore innovations in flow cytometry, imaging, and cell sorting. Whether you're looking to solve specific research challenges, streamline workflows, or experience the power of our latest technologies firsthand, this is your chance to engage directly with Thermo Fisher Scientific specialists.
Flow Cytometry User Group Meeting 2026
Watch on-demand sessions
Explore presentations from the Flow Cytometry User Group Meeting 2026 and discover the latest scientific insights, innovative applications, and expert perspectives shared by the flow cytometry community.
Don’t miss our tutorial sessions
Sunday, June 7 | 12:00–1:15 pm | Ballroom C
From discovery to sorting: an integrated spectral cytometry solution optimized by built-in tSNE/UMAP tools
Speaker: Brandon J. Trent, PhD
Abstract
Spectral cytometry has expanded high-parameter multicolor flow cytometry, enabling increasingly complex panels and deeper exploration of novel and rare immune cell populations. However, these advances place greater demands on instrument speed, acquisition efficiency, sorting performance, and data analysis. To address these challenges, we developed the Invitrogen Attune Xenith Flow Cytometer, which integrates spectral detection with proven acoustic focusing technology to deliver efficient, high-parameter sample acquisition at rates up to 1,000 µL/min while maintaining the resolution required to detect rare populations. Complementing this system, the Invitrogen Bigfoot Spectral Cell Sorter features a shared optical design and collection deck architecture to streamline panel transfer between platforms and incorporates innovative hardware to enable efficient up to six-way spectral sorting with high purity, efficiency, recovery, and throughput—helping provide a cohesive workflow solution from acquisition through sorting.
Using high-parameter immunophenotyping panels in mouse tissues and human peripheral blood mononuclear cells (PBMCs), we acquired data on the Attune Xenith Flow Cytometer and performed dimensionality reduction using tSNE or UMAP within Invitrogen Flowscape software. Flowscape utilizes automated identification and gating of populations of interest directly within UMAP space, facilitating rapid, data-driven population discovery. These analyses inform and optimize downstream sorting strategies on the Invitrogen Bigfoot Spectral Cell Sorter. Together, the Attune Xenith Flow Cytometer and Bigfoot Spectral Cell Sorter provide users with rapid detection, analysis, and high-purity isolation of rare immune cell subsets, supporting streamlined, high-dimensional flow cytometry workflows.
Tuesday, June 9 | 12:15–1:15 pm | Meeting Room 2F
From single cells to spatial landscapes: Integrating imaging-enhanced flow cytometry and spectral tissue imaging
Speakers: Nancie Mooney & Bethan Jones, PhD
Abstract
Cellular analysis is greatly enabled by spatial resolution by offering useful context at the cellular and tissue levels. This tutorial highlights how the Invitrogen Attune CytPix Flow Cytometer and the Invitrogen EVOS S1000 Spatial Imaging System help provide richer single cell-level information for better biological insights. While both technologies provide single-cell resolution, the Attune CytPix platform enables high throughput and AI-assisted management of imaging data and the EVOS S1000 system provides contextual information regarding the tissue microenvironment.
The Attune CytPix flow analyzer combines fluorescence, light scatter, and brightfield imaging to integrate quantitative morphometric features with phenotypic markers, enabling data interpretation in the context of cellular morphology and physiology. This utilization of Imaging-Enhanced Flow Cytometry (IEFC) strengthens confidence in cell identity and analytical decisions. User-trainable AI models allow researchers to tailor image analysis to their own samples, converting imaging data into robust, decision-ready metrics. Integrating morphology with fluorescence, alongside tools such as image similarity and dimensionality reduction, sharpens interpretation and strengthens analytical confidence.
Extending analysis beyond single-cell suspensions, the EVOS S1000 Spatial Imaging System delivers advanced multiplexed spectral imaging of intact tissue sections within a streamlined and accessible workflow. Multiple markers can be analyzed simultaneously on a single slide, with on-the-fly spectral unmixing that simplifies imaging and reduces post-acquisition processing. We will illustrate how multiplex spatial phenotyping reveals the localization and organization of immune and tumor cells, helping to provide critical context that cannot be obtained from flow cytometry alone.
Together, these technologies underscore how single-cell precision and spatial biology, whether from IEFC or multiplex fluorescence imaging, can be aligned to help provide complementary insights for a more complete understanding of complex biology.
Visit our scientific posters
Sunday, June 7, 2026 10:30 am–11:30 pm
Grand Ballroom
Jayanth Narayanan
Optimized toolset for evaluating single-color controls in spectral flow cytometry
Scientific oral presentation
Monday, June 8, 2026 5:00 pm–7:00 pm
Board P069
Susan O'Meara
Fast, efficient analysis of challenging and clog-prone tissue types using the Attune Xenith Flow Cytometer
Board P119
Dan Troast
Integrating post-unmixing compensation into spectral workflows
Board P187
Scott Sweeney
Reducing workflow burden in high-parameter cytometry with ready-to-use reagents
Board P203
Brandon Trent
From discovery to sorting: Integrated spectral analyzer and sorter solutions optimized by built-in tSNE/UMAP tools
Tuesday, June 9, 2026 5:00 pm–7:00 pm
Board P100
Mitch Klomp
Utilizing the 200 μm nozzle tip on the Bigfoot Spectral Cell Sorter to sort spheroids and tumoroids
Board P146
Adam York
Improvement of NIST fluorescence intensity assignments of calibration beads for quantitative flow cytometry
Board P166
Susan O’Meara
Jet-in-air vs. cuvette sorting: A modern comparison of post-sort viability across diverse cell types
Board P206
William Kang
NovaFluor dyes: Expanding multiparameter spectral flow cytometry
Featured innovations
Explore the latest technologies advancing cell analysis, imaging, and spectral flow cytometry.
From advanced cytometers to next-generation spatial imaging platforms, our latest innovations are engineered to accelerate discovery, simplify workflows, and deliver deeper insights. Whether you're designing high-parameter panels, integrating real-time imaging, or enhancing core lab performance, these solutions bring precision and power directly to your research.
See them live at Booth #213—experience what’s new, and what’s next.
Attune Xenith Flow Cytometer
High-performance spectral flow cytometry with acoustic-assisted focusing for fast, flexible, and reliable assay setup across conventional and spectral modes.
Attune CytPix Flow Cytometer
Integrates high-speed flow cytometry with real-time cellular imaging and AI-driven morphology analysis for deeper, data-rich insights.
Bigfoot Spectral Cell Sorter
High-throughput, high-parameter cell sorting with advanced spectral technology and a compact, ultra-quiet design built for core facility workflows.
EVOS S1000 Spatial Imaging System
Multiplex widefield imaging with intuitive software and single-round 9-plex spectral unmixing for streamlined, high-resolution spatial analysis.