Search
The sustainability and economic viability of some of society’s core bulk commodity processes relies on sorting, blending and controlling process streams with ever greater efficiency. Thermo Scientific CB Omni Agile Online Elemental Cross-Belt Analyzer delivers the real-time elemental analysis needed for success. Using market-leading Prompt Gamma Neutron Activation Analysis (PGNAA) technology or Pulsed Fast Thermo Neutron Activation (PFTNA) technology, it uniformly and precisely measures a wide range of industrially relevant elements, for entire raw material process streams. CB Omni Agile analyzer is lightweight, highly configurable, and easy to install. It allows miners and minerals processors, cement and steel manufacturers to reduce sampling and manual analysis, minimize environmental footprint, drive up process efficiency and boost profitability.
The CB Omni Agile Online Elemental Cross-Belt Analyzer is fully customizable with respect to key parameters such as neutron source, detector number, physical dimensions/geometry, and software. Our experience and expertise are crucial in using this flexibility to create a tailored solution for each application, one that will integrate within the existing plant or an evolving plant design, that will operate reliably, and that will help ensure the required accuracy and precision for your application.
PGNAA involves bombarding a sample with thermal neutrons and then measuring the prompt gamma rays released as excited nuclei return to their baseline state. A starting point is therefore the neutron source: either a radioactive isotope or a neutron generator.
Our technical experts have the freedom to configure the CB Omni Agile Analyzer with either Californium-252 (Cf252) or an electrically driven neutron generator depending on the material of interest and the specific application. Electrically driven neutron generators utilize the technique of PFTNA.
With PGNAA each element produces a prompt gamma ‘fingerprint’. The overall signal in a PGNAA measurement is therefore a function of the elements present and their relative abundance.
We use one, two or four large volume NaI scintillation gamma ray detectors for highly sensitive measurement depending on the application needs. We match detector(s) and source to deliver the accuracy and precision needed, given the elements present and their concentration.
The CB Omni Agile Analyzer has a lightweight modular design for easy installation and relocation. It fits all types of belts, including those that are steel corded, across a width range of 600 – 1800 mm. Tunnel height is adaptable to process needs. Innovative electronics and multiple options for plant connectivity facilitate integration with existing plant control hierarchies.
The real-time elemental data that the CB Omni Agile Analyzer provides a foundation for better manual control, automated control, and rapid upset detection, for a wide range of processes. The periodic table to the left, which shows ten-minute threshold of detectability data, highlights the elements that PGNAA is suitable for. Check those of interest for your process to assess its potential.
We understand that CB Omni Agile systems lie at the heart of operations that work to exacting onstream targets; unplanned downtime is prohibitively expensive.
Our global network of experts delivers services and support tailored to your business requirements. These plans control costs while at the same time simplifying in-house maintenance requirements. We are proactive, knowledgeable, experienced and responsive. Partner with us to keep your CB Omni Agile Online Elemental Cross-Belt Analyzer working reliably, 24/7, year after year with minimal effort.
With a service plan, you're covered in more ways than ever. You are the expert in your business, and we know how to keep your instrument running at top performance – together, we can build a partnership to achieve your production goals. Customers who partner with us for their service needs have access to our highest level of support and are investing in the longevity of their instrument. A service plan proactively protects instruments, maximizes uptime, manages yearly service expenditures, and allows you to focus on your core business.
For customers that want preferential treatment for their scheduled & unscheduled maintenance to limit interruptions.
For customers that want hassle-free service & corrective maintenance management with a predictable budget for their operations.
For customers that require support to keep their instruments at optimal performance and have limited technical staff availability.
|
Premium |
Advanced |
Preventive |
Preventive services |
|||
Preventive maintenance (PM) |
✓ |
✓ |
✓ |
Radiation inspection1,5 |
✓ |
✓ |
✓ |
Optimization & performance check1 |
✓ |
✓ |
✓ |
Static calibration2 |
✓ |
✓ |
10% off |
Dynamic calibration support2 |
✓ |
✓ |
10% off |
Operator training3 |
✓ |
✓ |
✓ |
Radiation services |
|||
Radiation source |
10% off (option to include) |
10% off (option to include) |
5% off |
Neutron generator repair6 |
10% off |
10% off |
5% off |
Installation and removal of radiation source1,6 |
✓ |
✓ |
✓ |
Disposal of the radiation source |
Option |
Option |
Option |
Corrective maintenance and instrument optimization |
|||
Remote technical support5 |
24/7 |
Business hours (Option-24/7) |
Business hours |
Targeted onsite response4 |
Top priority |
High priority |
Priority |
Labor & travel |
✓ |
✓ |
10% off |
Spare parts |
✓ |
10% off |
10% off |
System audit & consultancy |
Option |
Option |
Option |
Additional onsite application support |
10% off |
10% off |
|
Software upgrades |
✓ |
✓ |
|
System upgrades |
10% off |
10% off |
|
Thermo Scientific Omni View GUI is the primary user interface for the CB Omni Agile Online Elemental Cross-Belt Analyzer. Flexible and highly configurable, it allows multiple users to simultaneously access, process, and display data to meet individual and process requirements. With a multi-year database for data archiving and the ability to track system health, Omni View helps users to realize the full value that CB Omni Agile Analyzer can deliver.
Visit the PGNAA Technology Learning Center and browse our diverse range of documents, videos, webinars, and other resources below to see how you can cross-belt analyzers work for your specific application.
For Research Use Only. Not for use in diagnostic procedures.