Storage and Transport Solutions
Our portfolio of reusable support containers accommodates a range of functionalities and chamber dimensions. The interaction between the BPC and the outer container system is important in simple liquid-handling applications, where the BPC system is not shipped, and becomes even more critical when transportation takes place. We offer plastic and stainless steel options to suit your storage and transport needs.
Featured storage and transport solutions
Options to suit your storage and transport needs
Useful for internal and transportation applications. Top drain only and top- and bottom-drain, high-density polyethylene cylindrical drums from 50 L to 300 L, as well as square or rectangular polypropylene containers from 200 to 1,000 L are available. Regular and cylindrical tanks are also available in HDPE, LLPDE and PP in sizes ranging from 75 L to 757 L.
Thermo Scientific HyPerforma Smartainer 3.0 systems, with a range from 100 to 3,000 L. These containers are used for in-house, large-volume liquid-handling and storage operations.
Related product
Bioprocessing resources
Access a targeted collection of scientific application notes, case studies, posters, white papers and more for bioprocessing.
Explore our resources