Search
Catalog Number(s)
11126034| Components | Molecular Weight | Concentration (mg/L) | mM |
|---|---|---|---|
| Inorganic Salts | |||
| Calcium Chloride (CaCl2) (anhyd.) | 111.0 | 189.0 | 1.7027028 |
| Magnesium Sulfate (MgSO4-7H2O) | 246.0 | 294.0 | 1.195122 |
| Potassium Chloride (KCl) | 75.0 | 356.0 | 4.7466664 |
| Potassium Phosphate monobasic (KH2PO4) | 136.0 | 162.0 | 1.1911764 |
| Sodium Bicarbonate (NaHCO3) | 84.0 | 2106.0 | 25.071428 |
| Sodium Chloride (NaCl) | 58.0 | 5546.0 | 95.62069 |
| Other Components | |||
| Bovine Albumin Fraction V | 5000.0 | Infinity | |
| D-Glucose (Dextrose) | 180.0 | 1000.0 | 5.5555553 |
| DL-Lactic Acid Na | 112.06 | 2773.0 | 24.745672 |
| Sodium Pyruvate | 110.0 | 56.0 | 0.5090909 |