Search
| Components | Molecular Weight | Concentration (mg/L) | mM |
|---|---|---|---|
| Amino Acids | |||
| Glycine | 75.0 | 200.0 | 2.6666667 |
| Hydroxy L-proline | 131.0 | 800.0 | 6.10687 |
| L-Arginine hydrochloride | 211.0 | 800.0 | 3.791469 |
| L-Asparagine | 132.0 | 1300.0 | 9.848485 |
| L-Aspartic acid | 133.0 | 1300.0 | 9.774436 |
| L-Cystine-2Na | 286.0 | 119.14 | 0.41657344 |
| L-Glutamic Acid | 147.0 | 1500.0 | 10.204082 |
| L-Glutamine | 146.0 | 1000.0 | 6.849315 |
| L-Histidine | 155.0 | 200.0 | 1.2903225 |
| L-Isoleucine | 131.0 | 750.0 | 5.7251906 |
| L-Leucine | 131.0 | 250.0 | 1.908397 |
| L-Lysine hydrochloride | 183.0 | 700.0 | 3.8251367 |
| L-Methionine | 149.0 | 1000.0 | 6.7114096 |
| L-Phenylalanine | 165.0 | 1000.0 | 6.060606 |
| L-Proline | 115.0 | 500.0 | 4.347826 |
| L-Serine | 105.0 | 200.0 | 1.9047619 |
| L-Threonine | 119.0 | 200.0 | 1.6806723 |
| L-Tryptophan | 204.0 | 100.0 | 0.49019608 |
| L-Tyrosine disodium salt dihydrate | 261.0 | 360.4 | 1.3808429 |
| L-Valine | 117.0 | 500.0 | 4.2735043 |
| beta-Alanine | 89.0 | 300.0 | 3.3707864 |
| Vitamins | |||
| Biotin | 244.0 | 0.16 | 6.557377E-4 |
| Choline chloride | 140.0 | 20.0 | 0.14285715 |
| D-Calcium pantothenate | 477.0 | 0.008 | 1.677149E-5 |
| Folic Acid | 441.0 | 0.08 | 1.8140589E-4 |
| Nicotinic acid (Niacin) | 123.0 | 0.16 | 0.0013008129 |
| Para-Aminobenzoic Acid | 137.0 | 0.32 | 0.0023357663 |
| Pyridoxine hydrochloride | 206.0 | 0.4 | 0.0019417476 |
| Riboflavin | 376.0 | 0.08 | 2.1276595E-4 |
| Succinic acid | 118.0 | 4.8 | 0.04067797 |
| Thiamine hydrochloride | 337.0 | 0.08 | 2.3738873E-4 |
| Vitamin B12 | 1355.0 | 0.24 | 1.7712176E-4 |
| i-Inositol | 180.0 | 0.4 | 0.0022222223 |
| Inorganic Salts | |||
| Ammonium Molybdate ((NH4)6Mo7O24-4H2O) | 1236.0 | 0.04 | 3.236246E-5 |
| Calcium Chloride (CaCl2) (anhyd.) | 111.0 | 500.0 | 4.5045047 |
| Cobalt Chloride (CoCl2-6H2O) | 238.0 | 0.05 | 2.1008404E-4 |
| Cupric chloride (CuCl2-2H2O) | 170.0 | 0.2 | 0.0011764707 |
| Ferrous sulfate (FeSO4-7H2O) | 278.0 | 0.55 | 0.0019784174 |
| Magnesium Sulfate (MgSO4) (anhyd.) | 120.0 | 918.0 | 7.65 |
| Manganese chloride (MnCl2-4H2O) | 198.0 | 0.02 | 1.010101E-4 |
| Potassium Chloride (KCl) | 75.0 | 1200.0 | 16.0 |
| Sodium Bicarbonate (NaHCO3) | 84.0 | 350.0 | 4.1666665 |
| Sodium Chloride (NaCl) | 58.0 | 2850.0 | 49.13793 |
| Sodium Phosphate monobasic (NaH2PO4-H2O) | 138.0 | 1160.0 | 8.405797 |
| Zinc chloride (ZnCl2) | 136.0 | 0.04 | 2.9411764E-4 |
| Other Components | |||
| Alpha-Ketoglutaric acid | 146.0 | 29.6 | 0.20273973 |
| D-Glucose (Dextrose) | 180.0 | 2500.0 | 13.888889 |
| Fumaric acid | 116.0 | 4.4 | 0.037931036 |
| Malic acid | 134.0 | 53.6 | 0.39999998 |
| Maltose | 342.0 | 1000.0 | 2.9239767 |
| Sucrose | 342.0 | 1650.0 | 4.8245616 |
1. Schneider, I., (1964) Differentiation of larval Drosophila eye-antennal discs in vitro. J. Exp. Zool., 156:91.