The Role of Rapid NGS Testing in the MyeloMATCH Trial Initiative for AML and MDS
The Myeloid Malignancies Molecular Analysis for Therapy Choice, or MyeloMATCH, is an innovative umbrella trial designed to advance new treatment strategies for acute myeloid leukemia (AML) and myelodysplastic syndromes (MDS). This trial aims to enhance understanding and development of distinct protocols for patients based on their underlying genomic profiles.
In a recent webinar, Dr. Jerald Radich from the Fred Hutchinson Cancer Research Center emphasized the critical role of next-generation sequencing (NGS) technology in assessing disorders such as AML, which is essential for rapid identification and management.
Dr. Jerald Radich, MD
Member Clinical Research Division
Kurt Enslein Endowed Chair
Fred Hutchinson Cancer Research Center
The Importance of NGS in precision oncology
Physicians now have several therapeutic options available for AML patients, including multiple targeted therapies that have been approved in recent years. However, determining the most effective treatment strategy and therapy combination for a given patient is a complex task. By identifying specific mutations associated with a patient’s disease, clinicians can infer optimal therapeutic regimens for individuals based on their genomic profile, potentially improving patient outcomes.
MyeloMATCH trial sample screening and assessment
In the MyeloMATCH trial, AML patients will have samples drawn at one of the participating national clinical trial network (NCTN) sites. Within 72 hours of the molecular laboratory receiving the sample, it will undergo rapid NGS, flow cytometry, and genetic analysis to generate data for a Matchbox report. The NGS testing will be conducted using the Ion Torrent Genexus System, a comprehensive NGS solution that automates the entire workflow for the NCI Myeloid Assay.
The analysis will be carried out at the NCI Molecular Characterization Laboratory (MoCha) at the Frederick National Laboratory for Cancer Research or at Fred Hutchinson. Flow cytometry will be concurrently performed at Children’s Hospital Los Angeles and cytogenetics will be conducted at Fred Hutchinson Cancer Center. Additionally, a significant number of samples will be sent to nationwide repositories for future research.
This combination of tests, representing the infrastructure of this trial, received Investigational Device Exemption (IDE) approval from the Food and Drug Administration (FDA). The NCI Myeloid Assay and the Genexus System were included in the IDE approval.
72-hour study assignment
The data from the trial will inform a Matchbox report, an algorithm-driven approach based on multiple variables that will help assign the patient to a sub-study within 72 hours. This rapid process is crucial due to the urgency typically required for treating diseases like AML, where prompt treatment initiation can lead to better prognoses.
Summary
The MyeloMATCH trial, with its strategic application of NGS, is at the forefront of precision oncology for AML and MDS. The insights from the trial are driving the field towards more accurate and efficient diagnostic and therapeutic approaches. Several clinical sites are already open, with more to come. As NGS technology continues to evolve, our ability to manage and treat these complex diseases with unprecedented precision will also advance, thanks in part to studies such as MyeloMATCH and future technological innovations.
Ready to speak to a Thermo Fisher representative?
We will be happy to answer your questions and provide a demo of our NGS solutions.
For Research Use Only. Not for use in diagnostic procedures.
CN: 60360