Options and accessories for any application and lab environment.
Heratherm Environmental Chambers give you reliable, precise control over a wide range of temperature, humidity, and lighting requirements. Designed to be versatile, our chambers provide ideal environments across product and packaging stability testing, plant growth, and animal hatching applications.
From stability and incubation chambers to light chambers, versatile functionality comes in a sophisticated and easy-to-use package that helps you achieve reliable results.
Modular design and extensive options to create your ideal chamber.
All Heratherm Environmental Chamber models are built to be dependable fixtures in your lab, giving you precise control of environmental factors. The thoughtful design and flexible options make them even more versatile, giving you the flexibility to order a configuration that perfectly suits your application and lab environment.
Door Options
Choose between a glass door or a solid door with inner glass door to support application requirements. Additional door options include left-or-right open and door lock. 
Cabinet Options
Cabinet options include: Duplex electrical sockets inside the chamber; a water connection to a clean water line can replace the internal water tank; and two standard access ports cover most needs for use of sensors, probes and accessory cables inside the chamber. Please contact us for custom access port options
Shelving
Choose from solid stainless steel, reinforced, or perforated stainless steel shelving depending on your intended use and accommodating from 30 kg / 66.1 lbs per shelf. Reinforced perforated stainless steel shelves carry up to 90 Kg / 198.4 lbs. An optional reinforced shaker shelf system increase stability and quiet operation of the chamber. 
Integrated light options
Light chambers come with programmable range of light color and intensity, with a choice of up to three levels of light exposure. Select between two light module options: light module for ICH, with white LED lights, and UV LED light, or light module for plant/animal growth with white LED lights.