Search
Refrigerated incubator chambers meet International Conference on Harmonization (ICH) guidelines for drug substance and product storage testing. The units are ideal for drug stability studies, shelf-life testing for packaged products, and other large-scale biological research.
Temperature
0°C to 60°C (32°F to 140°F) temperature range that can be set in 0.1°C increments.
Stability and uniformity
Optimum temperature uniformity and recovery contribute to an ideal test environment.
Large capacity
Accommodates large product loads and low-output, heat-generating equipment.
|
Refrigerated Reach-In Chamber |
Size |
29 cu. ft. |
Exterior (W x H x D) |
965 x 2032 x 833mm |
Interior (W x H x D) |
787 x 1524 x 686mm |
Net weight |
318kg / 700 lbs |
Shipping weight |
386kg / 850 lbs |
Temperature |
|
Temperature range |
0°C to 60°C |
Controller, sensor |
Watlow Microprocessor - PID, with RTD sensor |
Display |
Digital dual LED, 0.1°C steps readability & setability |
Temperature Uniformity |
≤ ±0.3°C @ 20° to 37°C |
Temperature Stability |
@ 20° to 45°C |
Electrical |
|
Voltage |
120V, 60Hz, 16.0 Amps 3 Wire |
Plug |
NEMA 5-20P |
Power switch |
2-Pole |
Interior Convenience receptacles |
Optional |
Alarm Contacts |
Temp. deviation and power failure; Common, NO, NC |
Available Options |
|
Order Number |
|