B-27™ Supplement (50X), custom - FAQs
B-27™ Supplement (50X), custom - FAQs
View additional product information for B-27™ Supplement (50X), custom - FAQs (0080085SA)
1 product FAQs found
Do you offer B-27 supplements with and without insulin?
Yes, we do offer B-27 Supplement, minus insulin (Cat. Nos. A1895601 and A1895602). All our other B-27 supplements do contain insulin.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.