GeneTitan™ MC Xenon Lamp - FAQs
GeneTitan™ MC Xenon Lamp - FAQs
View additional product information for GeneTitan™ MC Xenon Lamp - FAQs (01-0740)
1 product FAQs found
What is the catalog number for the GeneTitan MC Xenon Lamp?
The catalog number for the GeneTitan MC Xenon Lamp is 01-0740.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.