UltraComp eBeads™ Plus Compensation Beads, 25 tests - FAQs
View additional product information for UltraComp eBeads™ Plus Compensation Beads - FAQs (01-3333-41, 01-3333-42)
1 product FAQs found
What is the size of UltraComp eBeads Plus Compensation Beads?
The bead size is 5 µm.