Tungsten foil, 0.05mm (0.002in) thick, 99.95% (metals basis) - FAQs
Tungsten foil, 0.05mm (0.002in) thick, 99.95% (metals basis) - FAQs
View additional product information for Tungsten foil, 0.05mm (0.002in) thick, 99.95% (metals basis) - FAQs (010417.GH, 010417.FI, 010417.HP)
1 product FAQs found
Is there a coating on the Tungsten foil, 0.05mm (0.002in) thick, 99.95% (metals basis)?
The Tungsten foil, 0.05mm (0.002in) thick, 99.95% (metals basis) may have an oil or oxide coating. To remove the coating, we recommend soaking the foil in very hot water with strong detergent for 10-15 minutes. Wipe the foil with a sponge. Then rinse it with water and dry. This process will remove the coating.