Zinc rod, 1.27cm (0.5in) dia, 99.99+% (metals basis)
Zinc rod, 1.27cm (0.5in) dia, 99.99+% (metals basis)
Zinc rod, 1.27cm (0.5in) dia, 99.99+% (metals basis)
Thermo Scientific Chemicals

Zinc rod, 1.27cm (0.5in) dia, 99.99+% (metals basis)

CAS: 7440-66-6 | Zn | 65.38 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
30 cm
60 cm
90 cm
Catalog number 010433.BV
also known as 010433-BV
Price (USD)
96.65
Online exclusive
107.00
Save 10.35
Each
Quantity:
30 cm
Request bulk or custom format
Price (USD)
96.65
Online exclusive
107.00
Save 10.35
Each
Chemical Identifiers
CAS7440-66-6
IUPAC Namezinc
Molecular FormulaZn
InChI KeyHCHKCACWOHOZIP-UHFFFAOYSA-N
SMILES[Zn]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White-silver
FormRod, 1.27cm (0.5in) diameter.
Assay from Supplier's CofA>99.99% (metals basis)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only