Zinc shot, 1-6mm (0.04-0.24in), Puratronic™, 99.9999% (metals basis)
Zinc shot, 1-6mm (0.04-0.24in), Puratronic™, 99.9999% (metals basis)
Zinc shot, 1-6mm (0.04-0.24in), Puratronic™, 99.9999% (metals basis)
Zinc shot, 1-6mm (0.04-0.24in), Puratronic™, 99.9999% (metals basis)
Thermo Scientific Chemicals

Zinc shot, 1-6mm (0.04-0.24in), Puratronic™, 99.9999% (metals basis)

CAS: 7440-66-6 | Zn | 65.38 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1 kg
Catalog number 010760.A1
also known as 010760-A1
Price (USD)
2,724.65
Online Exclusive
3,027.00
Save 302.35 (10%)
Each
Quantity:
1 kg
Request bulk or custom format
Price (USD)
2,724.65
Online Exclusive
3,027.00
Save 302.35 (10%)
Each
Chemical Identifiers
CAS7440-66-6
IUPAC Namezinc
Molecular FormulaZn
InChI KeyHCHKCACWOHOZIP-UHFFFAOYSA-N
SMILES[Zn]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.0001% max.

Polytetrafluoroethylene, 6-10 micron is used in specially designed coating applications on coil for bakeware.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

RUO – Research Use Only