Zinc rod, 10mm (0.39in) dia, 99.9998% (metals basis)
Zinc rod, 10mm (0.39in) dia, 99.9998% (metals basis)
Zinc rod, 10mm (0.39in) dia, 99.9998% (metals basis)
Thermo Scientific Chemicals

Zinc rod, 10mm (0.39in) dia, 99.9998% (metals basis)

CAS: 7440-66-6 | Zn | 65.38 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mm
250 mm
Catalog number 012718.DT
also known as 012718-DT
Price (USD)
445.65
Special offer
Online exclusive
Ends: 30-Jun-2026
524.00
Save 78.35 (15%)
Each
Quantity:
250 mm
Request bulk or custom format
Price (USD)
445.65
Special offer
Online exclusive
Ends: 30-Jun-2026
524.00
Save 78.35 (15%)
Each
Specifications
Assay Percent Notes(metals basis)
CAS7440-66-6
ChEBICHEBI:30185
Chemical Name or MaterialZinc rod
EINECS Number231-175-3
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only