Yttrium Aluminum lump, 99.9% (metals basis excluding Ca), Ca 1% max
Thermo Scientific Chemicals

Yttrium Aluminum lump, 99.9% (metals basis excluding Ca), Ca 1% max

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 012949.14
also known as 012949-14
Price (USD)
102.65
Special offer
Online exclusive
Ends: 28-Dec-2026
121.00
Save 18.35
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
102.65
Special offer
Online exclusive
Ends: 28-Dec-2026
121.00
Save 18.35
Each
Specifications
Assay Percent Notes(metals basis excluding Ca), Ca 1% max.
Avg. Mol. Wt. or Mol. Wt. RangeY:Al; 87:12 wt%
Chemical Name or MaterialYttrium Aluminum lump
MDL NumberMFCD01074900
Recommended StorageAmbient temperatures
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only