What is the maximum working temperature of Al-23 Tubes?
The maximum working temperature for the Al-23 Tube is 1950 degrees C.
Search
View additional product information for Al-23 Tube (both ends open);OD (mm), 30;ID (mm), 25 - FAQs (031574.DR, 031574.EH)
1 product FAQs found