What is the maximum working temperature of Al-23 Tubes?
The maximum working temperature for the Al-23 Tube is 1950 degrees C.
Search
View additional product information for Al-23 Fish Spine Beads;OD (mm), 3.5;Bore (mm), 1.0;Length (mm), 3.0 - FAQs (032905.KX)
1 product FAQs found