Al-23 Crucible, Tammann;Height x OD x ID (mm), 50 x 8 x 5;Volume (ml), 1;Tammann - FAQs
View additional product information for Al-23 Crucible, Tammann;Height x OD x ID (mm), 100 x 12 x 8;Volume (ml), 5;Tammann - FAQs (033053.KF, 033055.KF)
1 product FAQs found
What is the maximum working temperature of Al-23 Tubes?
The maximum working temperature for the Al-23 Tube is 1950 degrees C.