D-Mannitol, ACS
D-Mannitol, ACS
D-Mannitol, ACS
Thermo Scientific Chemicals

D-Mannitol, ACS

CAS: 69-65-8 | C6H14O6 | 182.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2.5 kg
Catalog number 033342.36
also known as 033342-36
Price (USD)
75.65
Online exclusive
84.30
Save 8.65
Each
Quantity:
500 g
Request bulk or custom format
Price (USD)
75.65
Online exclusive
84.30
Save 8.65
Each
Chemical Identifiers
CAS69-65-8
IUPAC Namehexane-1,2,3,4,5,6-hexol
Molecular FormulaC6H14O6
InChI KeyFBPFZTCFMRRESA-UHFFFAOYNA-N
SMILESOCC(O)C(O)C(O)C(O)CO
View more
SpecificationsSpecification SheetSpecification Sheet
Impurity contentReducing sugars P.T.
Free acid (titration)0.0008meq/g
Optical RotationA25/D +23.3? to +24.3?
Insoluble material0.01%
Trace Metals(as Pb) 5ppm
View more
D-Mannitol is used with boric acid to make dry electrolytic condensers for radio applications, artificial resins and plasticizers. It is used as excipient for chewable tablets due to its pleasant taste. It is used in volumetric analysis of boric acid in analytical chemistry. It is also used in the food industry as anticaking, free-flow agent, flavoring agent, stabilizer, thickener and nutritive sweetener. Further, it is used in medicine as diuretic and in the determination of renal function. In addition, it is used as a formulation aid, lubricant, non-nutritive sweetener and solvent for foods.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only