Lanthanum rod, 6.35mm (0.25in) dia, 99.9% (REO)
Lanthanum rod, 6.35mm (0.25in) dia, 99.9% (REO)
Lanthanum rod, 6.35mm (0.25in) dia, 99.9% (REO)
Thermo Scientific Chemicals

Lanthanum rod, 6.35mm (0.25in) dia, 99.9% (REO)

Form: Approx.0.20g/mm, Cast rod, diameter is approximate | CAS: 7439-91-0 | La | 138.91 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 mm
25 mm
50 mm
100 mm
Catalog number 040292.DH
also known as 040292-DH
Price (USD)
212.65
Special offer
Online exclusive
Ends: 30-Jun-2026
250.00
Save 37.35 (15%)
Each
Quantity:
25 mm
Request bulk or custom format
Price (USD)
212.65
Special offer
Online exclusive
Ends: 30-Jun-2026
250.00
Save 37.35 (15%)
Each
Chemical Identifiers
CAS7439-91-0
IUPAC Namelanthanum
Molecular FormulaLa
InChI KeyFZLIPJUXYLNCLC-UHFFFAOYSA-N
SMILES[La]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.1% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only