Lanthanum rod, 6.35mm (0.25in) dia, 99.9% (REO)
Lanthanum rod, 6.35mm (0.25in) dia, 99.9% (REO)
Lanthanum rod, 6.35mm (0.25in) dia, 99.9% (REO)
Thermo Scientific Chemicals

Lanthanum rod, 6.35mm (0.25in) dia, 99.9% (REO)

Form: Approx.0.20g/mm, Cast rod, diameter is approximate | CAS: 7439-91-0 | La | 138.91 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 mm
25 mm
50 mm
100 mm
Catalog number 040292.DM
also known as 040292-DM
Price (USD)
791.65
Online Exclusive
879.00
Save 87.35 (10%)
Each
Quantity:
100 mm
Request bulk or custom format
Price (USD)
791.65
Online Exclusive
879.00
Save 87.35 (10%)
Each
Chemical Identifiers
CAS7439-91-0
IUPAC Namelanthanum
Molecular FormulaLa
InChI KeyFZLIPJUXYLNCLC-UHFFFAOYSA-N
SMILES[La]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.1% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only