Yttrium rod, 6.35mm (0.25in) dia, 99.9% (metals basis excluding Ta)
Yttrium rod, 6.35mm (0.25in) dia, 99.9% (metals basis excluding Ta)
Yttrium rod, 6.35mm (0.25in) dia, 99.9% (metals basis excluding Ta)
Thermo Scientific Chemicals

Yttrium rod, 6.35mm (0.25in) dia, 99.9% (metals basis excluding Ta)

CAS: 7440-65-5 | Y | 88.91 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 mm
25 mm
50 mm
100 mm
Catalog number 040299.DJ
also known as 040299-DJ
Price (USD)
391.65
Online Exclusive
435.00
Save 43.35 (10%)
Each
Quantity:
50 mm
Request bulk or custom format
Price (USD)
391.65
Online Exclusive
435.00
Save 43.35 (10%)
Each
Chemical Identifiers
CAS7440-65-5
IUPAC Nameyttrium
Molecular FormulaY
InChI KeyVWQVUPCCIRVNHF-UHFFFAOYSA-N
SMILES[Y]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities excluding Ta0.1% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Packaged under argon
RUO – Research Use Only