Search
View additional product information for Polyvinyl alcohol, 98-99% hydrolyzed, high molecular weight - FAQs (041243.22, 041243.14, 041243.36)
1 product FAQs found
Polyvinyl alcohol, 98-99% hydrolyzed, high molecular weight has a typical average molecular weight of 126,000 g/mol.