Lanthanum foil, 0.1mm (0.004 in.) thick, 99.9% (REO), Thermo Scientific Chemicals
Lanthanum foil, 0.1mm (0.004 in.) thick, 99.9% (REO), Thermo Scientific Chemicals
Lanthanum foil, 0.1mm (0.004 in.) thick, 99.9% (REO), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Lanthanum foil, 0.1mm (0.004 in.) thick, 99.9% (REO), Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
041558.FF
also known as 041558-FF
25 x 25 mm
Catalog number 041558.FF
also known as 041558-FF
Price (USD)
-
Quantity:
25 x 25 mm
Request bulk or custom format
Chemical Identifiers
CAS7439-91-0
IUPAC Namelanthanum
Molecular FormulaLa
InChI KeyFZLIPJUXYLNCLC-UHFFFAOYSA-N
SMILES[La]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.1% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Packaged under argon
RUO – Research Use Only