Chromium cubes, typically 6.35mm (0.25in) square, 99.95% (metals basis)
Chromium cubes, typically 6.35mm (0.25in) square, 99.95% (metals basis)
Chromium cubes, typically 6.35mm (0.25in) square, 99.95% (metals basis)
Thermo Scientific Chemicals

Chromium cubes, typically 6.35mm (0.25in) square, 99.95% (metals basis)

CAS: 7440-47-3 | Cr | 52.00 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number 042310.18
also known as 042310-18
Price (USD)
156.65
Special offer
Online exclusive
Ends: 30-Jun-2026
184.00
Save 27.35 (15%)
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
156.65
Special offer
Online exclusive
Ends: 30-Jun-2026
184.00
Save 27.35 (15%)
Each
Chemical Identifiers
CAS7440-47-3
IUPAC Namechromium
Molecular FormulaCr
InChI KeyVYZAMTAEIAYCRO-UHFFFAOYSA-N
SMILES[Cr]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Gray
Assay from Supplier's CofA≥99.95% (metals basis)
Formcube, typically 6.35mm (0.25in) square.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only