Zinc wire, 2.0mm (0.08in) dia, Puratronic™, 99.999% (metals basis)
Zinc wire, 2.0mm (0.08in) dia, Puratronic™, 99.999% (metals basis)
Zinc wire, 2.0mm (0.08in) dia, Puratronic™, 99.999% (metals basis)
Thermo Scientific Chemicals

Zinc wire, 2.0mm (0.08in) dia, Puratronic™, 99.999% (metals basis)

Form: approx. 22.4g/m | CAS: 7440-66-6 | Zn | 65.38 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 cm
100 cm
Catalog number 042637.BU
also known as 042637-BU
Price (USD)
108.65
Online Exclusive
121.00
Save 12.35 (10%)
Each
Quantity:
25 cm
Request bulk or custom format
Price (USD)
108.65
Online Exclusive
121.00
Save 12.35 (10%)
Each
Specifications
AppearanceGray wire
Assay Percent Range99.999% (Metals basis)
CAS7440-66-6
ChEBICHEBI:30185
Chemical Name or MaterialZinc
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only