Chromium granules, -4+7 mesh, 99.999% (metals basis)
Chromium granules, -4+7 mesh, 99.999% (metals basis)
Chromium granules, -4+7 mesh, 99.999% (metals basis)
Thermo Scientific Chemicals

Chromium granules, -4+7 mesh, 99.999% (metals basis)

CAS: 7440-47-3 | Cr | 52.00 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number 042926.30
also known as 042926-30
Price (USD)
1,377.65
Online Exclusive
1,530.00
Save 152.35 (10%)
Each
Quantity:
250 g
Request bulk or custom format
Price (USD)
1,377.65
Online Exclusive
1,530.00
Save 152.35 (10%)
Each
Chemical Identifiers
CAS7440-47-3
IUPAC Namechromium
Molecular FormulaCr
InChI KeyVYZAMTAEIAYCRO-UHFFFAOYSA-N
SMILES[Cr]
View more
SpecificationsSpecification SheetSpecification Sheet
FormGranules,
Particle Size-4+7 mesh
Assay from Supplier's CofA≥99.998% metals basis
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only