Methyl cellulose, viscosity 1600 cPs
Methyl cellulose, viscosity 1600 cPs
Methyl cellulose, viscosity 1600 cPs
Thermo Scientific Chemicals

Methyl cellulose, viscosity 1600 cPs

CAS: 9004-67-5 | C20H38O11 | 454.513 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number 043146.36
also known as 043146-36
Price (USD)
140.00
Each
Quantity:
500 g
Request bulk or custom format
Price (USD)
140.00
Each
Chemical Identifiers
CAS9004-67-5
IUPAC Name3,4,5-trimethoxy-2-(methoxymethyl)-6-{[4,5,6-trimethoxy-2-(methoxymethyl)oxan-3-yl]oxy}oxane
Molecular FormulaC20H38O11
InChI KeyYLGXILFCIXHCMC-UHFFFAOYNA-N
SMILESCOCC1OC(OC2C(COC)OC(OC)C(OC)C2OC)C(OC)C(OC)C1OC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream or pale yellow
Assay from Supplier's CofAViscosity: 1000-1800 mPa-s (2% in watre,20°C)
Sulfated Ash≤1.5%
FormPowder
Loss on Drying≤8.0%
Thickener and emulsifier in various food and cosmetic products, and also as a treatment of constipation. Methyl cellulose, as a gel, has the unique property of setting when hot and melting when cold. Methyl cellulose is used as a variable viscosity personal lubricant. Methyl cellulose is used in the manufacture of capsules in nutritional supplements. Methyl cellulose finds a major application as a performance additive in construction materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only