Methyl cellulose, viscosity 400 cPs
Methyl cellulose, viscosity 400 cPs
Methyl cellulose, viscosity 400 cPs
Thermo Scientific Chemicals

Methyl cellulose, viscosity 400 cPs

CAS: 9004-67-5 | C20H38O11 | 454.513 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Catalog number 043147.A4
also known as 043147-A4
Price (USD)
490.00
Each
Quantity:
2.5 kg
Request bulk or custom format
Price (USD)
490.00
Each
Chemical Identifiers
CAS9004-67-5
IUPAC Name3,4,5-trimethoxy-2-(methoxymethyl)-6-{[4,5,6-trimethoxy-2-(methoxymethyl)oxan-3-yl]oxy}oxane
Molecular FormulaC20H38O11
InChI KeyYLGXILFCIXHCMC-UHFFFAOYNA-N
SMILESCOCC1OC(OC2C(COC)OC(OC)C(OC)C2OC)C(OC)C(OC)C1OC
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Viscosity300-560 mPas/cPs (2% solution, 20°C)
Appearance (Color)White to grey
Residue on ignition≤2.0%
Assay from Suppliers CofAAssay, dried basis (methoxy groups): 26.0-33.0%
View more
Methyl cellulose is used as a thickener and emulsifier in various food and cosmetic products like hair shampoos, toothpastes and liquid soaps. It is used in the preparation of capsules in nutritional supplements. It is also used as a gel in advanced cookery and as a lubricant. It is also involved in the treatment of constipation. It acts as a buffer additive in capillary electrophoresis to control electoosmotic flow for improved separations. Further, it is used as a performance additive in construction materials and as sizing in the production of papers and textiles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only