Zinc foil, 2mm (0.08in) thick, Puratronic™, 99.999% (metals basis)
Zinc foil, 2mm (0.08in) thick, Puratronic™, 99.999% (metals basis)
Zinc foil, 2mm (0.08in) thick, Puratronic™, 99.999% (metals basis)
Thermo Scientific Chemicals

Zinc foil, 2mm (0.08in) thick, Puratronic™, 99.999% (metals basis)

CAS: 7440-66-6 | Zn | 65.38 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 x 25 mm
50 x 50 mm
Catalog number 043504.FF
also known as 043504-FF
Price (USD)
442.65
Special offer
Online exclusive
Ends: 30-Jun-2026
520.00
Save 77.35 (15%)
Each
Quantity:
25 x 25 mm
Request bulk or custom format
Price (USD)
442.65
Special offer
Online exclusive
Ends: 30-Jun-2026
520.00
Save 77.35 (15%)
Each
Specifications
Assay Percent Notes(metals basis)
CAS7440-66-6
ChEBICHEBI:30185
Chemical Name or MaterialZinc foil
EINECS Number231-175-3
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only