Zinc rod, 3.18mm (0.125in) dia, 99.994% (metals basis)
Zinc rod, 3.18mm (0.125in) dia, 99.994% (metals basis)
Zinc rod, 3.18mm (0.125in) dia, 99.994% (metals basis)
Thermo Scientific Chemicals

Zinc rod, 3.18mm (0.125in) dia, 99.994% (metals basis)

CAS: 7440-66-6 | Zn | 65.38 g/mol
Have Questions?
Catalog NumberQuantity
043695.HO350 mm
Catalog number 043695.HO
Price (USD)
158.65
Special offer
Online exclusive
Ends: 30-Jun-2026
186.00
Save 27.35 (15%)
Each
Quantity:
350 mm
Request bulk or custom format
Price (USD)
158.65
Special offer
Online exclusive
Ends: 30-Jun-2026
186.00
Save 27.35 (15%)
Each
Chemical Identifiers
CAS7440-66-6
IUPAC Namezinc
Molecular FormulaZn
InChI KeyHCHKCACWOHOZIP-UHFFFAOYSA-N
SMILES[Zn]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White silver
FormRod, 3.18mm (0.125in) diameter.
Assay from Supplier's CofA≥99.994% (metals basis)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only