Zinc Copper composite, -100 mesh, 99% (metals basis)
Thermo Scientific Chemicals

Zinc Copper composite, -100 mesh, 99% (metals basis)

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
100 g
500 g
Catalog number 044013.09
also known as 044013-09
Price (USD)
30.60
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
30.60
Each
Specifications
Assay Percent Notes(metals basis)
Avg. Mol. Wt. or Mol. Wt. RangeTypically Zn 90 to 95, Cu 5 to 10 wt%
Chemical Name or MaterialZinc Copper composite
DOT InformationTransport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL POWDERS, FLAMMABLE, N.O.S.
Health Hazard 1H228
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only