2-Aminoadenosine, 97%
Thermo Scientific Chemicals

2-Aminoadenosine, 97%

CAS: 2096-10-8 | C10H14N6O4 | 282.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number 044748.03
also known as 044748-03
Price (USD)
46.65
Special offer
Online exclusive
Ends: 30-Jun-2026
54.60
Save 7.95 (15%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
46.65
Special offer
Online exclusive
Ends: 30-Jun-2026
54.60
Save 7.95 (15%)
Each
Chemical Identifiers
CAS2096-10-8
IUPAC Name(2R,3R,4S,5R)-2-(2,6-diamino-9H-purin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol
Molecular FormulaC10H14N6O4
InChI KeyZDTFMPXQUSBYRL-SBTUYZBDNA-N
SMILESNC1=NC(N)=C2N=CN([C@@H]3O[C@H](CO)[C@@H](O)[C@H]3O)C2=N1
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Appearance (Color)White
Assay from Supplier's CofA≥96.0% (HPLC)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only