Dionex™ CR-TC Continuously Regenerated Trap Columns
Actual product may vary
Dionex™ CR-TC Continuously Regenerated Trap Columns
Dionex™ CR-TC Continuously Regenerated Trap Columns
Thermo Scientific™

Dionex™ CR-TC Continuously Regenerated Trap Columns

Remove ionic contaminants from eluents continuously without offline chemical regeneration using Dionex CR-TC Continuously Regenerated Trap columns.

Have Questions?
Change viewbuttonViewtableView
Catalog NumberColumn TypeFor Use With (Equipment)
060477Anion Trap ColumnStandard Anion Capillary RFIC Systems up to 3,000 psi
072078Anion Trap ColumnStandard Anion Capillary RFIC Systems
075550Anion Trap ColumnAll Anion, RFIC, and HPIC Systems that are Not Consumables Monitoring Enabled
066262Cation Trap ColumnStandard RFIC Systems up to 3,000 psi
072079Cation Trap ColumnStandard Cation Capillary RFIC Systems
088662Anion Trap ColumnDionex ICS-6000, Dionex Integrion, and Dionex Inuvion Systems
088663Cation Trap ColumnDionex ICS-6000, Dionex Integrion, and Dionex Inuvion Systems
075551Cation Trap ColumnDionex HPIC Systems that are Not Consumables Monitoring Enabled
104-60001Cation Trap ColumnDionex ICS-6000, Dionex Integrion, and Dionex Inuvion Systems
Catalog number 060477
Price (USD)
1,820.00
Each 1
Column Type:
Anion Trap Column
For Use With (Equipment):
Standard Anion Capillary RFIC Systems up to 3,000 psi
Price (USD)
1,820.00
Each 1

Remove ionic contaminants from eluents continuously, without the need for offline chemical regeneration. Thermo Scientific™ Dionex™ CR-TC Continuously Regenerated Trap Columns are electrolytically regenerated trap columns specifically designed for Reagent-Free™ ion chromatography (RFIC™) systems. When plumbed after the Thermo Scientific™ Dionex™ EGC Eluent Generation Cartridge, the Dionex CR-TC trap column removes all ionic contaminants from the deionized feed water, providing low drift during gradient operations.

Meet Today's Demands in Ion Chromatography

Dionex CR-TC columns:

  • Increase productivity
  • Provide very low baseline drift for improved integration and increased sensitivity
  • Provide contaminant-free deionized source water and eluents

Which Dionex CR-TC column to use with which IC system:

  • Use Dionex CR-ATC and CR-CTC II columns on standard RFIC systems that operate up to 3,000 psi
  • Use Dionex CR-TC 500 columns on all RFIC and high-pressure IC (HPIC) systems that operate up to 5,000 psi. You can use the Dionex CR-TC 500 column in place of the Dionex CR-ATC and CR-CTC II column
  • Use capillary Dionex CR-TC columns on capillary RFIC systems; do not use capillary Dionex CR-TC columns on analytical-scale RFIC systems
  • Use the Dionex CR-TC 600 and CR-CTC columns on the Thermo Scientific™ Dionex™ Inuvion™, Thermo Scientific™ Dionex™ Integrion™ and Thermo Scientific™ Dionex™ ICS-6000 HPIC Systems only

Remove Cationic Contaminants with Dionex CR-CTC III Columns

Dionex CR-CTC III Continuously Regenerated Cation Trap columns are high-pressure electrolytically regenerated trap columns designed for use with eluent generator devices and compatible with Dionex Inuvion, Dionex Integrion and Dionex ICS-6000 HPIC systems

The Dionex CR-CTC III offers:

  • Contaminant-free deionized source water and eluent
  • Time savings; no need to perform regeneration offline
  • Very low baseline drift for improved integration and increased sensitivity
  • Increased productivity; quality data soon after startup
  • Removal of cationic impurities from source water
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Crosslinking8% DVB
DescriptionDionex™ CR-ATC Continuously Regenerated Anion Trap Column
For Use With (Application)Removal of Ionic Contaminants from Eluents Without the Need for Off-Line Chemical Regeneration
For Use With (Equipment)Standard Anion Capillary RFIC Systems up to 3,000 psi
HydrophobicityLow
Max. Pressure3000 psi (207 bar)
Particle Size55 μm
QuantityEach 1
Void Volume (Metric)<100 μL
Column TypeAnion Trap Column
Unit SizeEach 1