Dionex™ IonPac™ Anion Analyte Trap Column
Dionex™ IonPac™ Anion Analyte Trap Column
Thermo Scientific™
Dionex™ IonPac™ Anion Analyte Trap Column
The Thermo Scientific™ Dionex IonPac Anion Analyte Trap Column will remove all the analyte anions from a sample in RFIC systems with Eluent Regeneration.
Have Questions?
Catalog number 064924
Price (MXN)
-
Request A Quote
The Thermo Scientific™ Dionex IonPac Anion Analyte Trap Column will remove all the analyte anions from a sample in RFIC systems with Eluent Regeneration.
Specifications
For Use With (Application)Removal of Analyte Anions from Samples via Eluent Regeneration
For Use With (Equipment)RFIC Systems
QuantityEach 1
Column TypeTrap Column
Unit SizeEach 1