B·R·A·H·M·S™ AFP KRYPTOR™
Thermo Scientific™
B·R·A·H·M·S™ AFP KRYPTOR™
B·R·A·H·M·S provides a complete portfolio of biomarkers for the most reliable results in prenatal screening. Reach high precision for trisomy screening in first and second trimester of pregnancy and neural tube defect screening in second trimester of pregnancy with B·R·A·H·M·S AFP KRYPTOR.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoTipo
081691NCalibrador
816075NEnsayos de inmunofluorescencia
081693NDiluyente
Número de catálogo 081691N
Precio (MXN)
-
Tipo:
Calibrador
B·R·A·H·M·S provides a complete portfolio of biomarkers for the screening of fetal defects in first and second trimester of pregnancy and biomarkers for diagnosis and monitoring of cancer. B·R·A·H·M·S AFP KRYPTOR is indicated as an aid to be used in conjunction with clinical evaluation to help identify pregnancies at risk of trisomy 21 in first trimester, of trisomy 21 and 18 and neural tube defects in second trimester, and as an aid for the diagnosis and monitoring of patients with hepatocellular carcinomas or non-seminomatous germ cell tumors.
US Only: Product is not 510k cleared in US.
Get confidence in your results with the high precision of B·R·A·H·M·S AFP KRYPTOR and its short incubation time.
  • Save cost and time with a multiparametric control containing 3 parameters: PAPP-A, Free βhCG and AFP
  • Incubation time of only 9 minutes for a short turnaround time
  • Allows measurement in serum, plasma (heparin) and amniotic fluid
  • Large capacity with 75 tests
  • Kit stable up to 15 days after opening onboard the instrument
  • Easy workflow with only 1 calibrator and one calibration every 2 weeks
  • Fits your throughput need with B·R·A·H·M·S KRYPTOR compact PLUS or B·R·A·H·M·S KRYPTOR GOLD
  • Allows risk calculation in first and second trimester with B·R·A·H·M·S Fast screen pre I plus
Thermo Fisher Scientific products are distributed globally and their uses, applications, indications, claims and availability of products in each country depend on local regulatory marketing authorization status, please consult the Instructions For Use (IFU) available in your country.
Especificaciones
Tiempo de análisis9 minutes
DescripciónCalibrator (1 level)
Para utilizar con (aplicación)Readjust the standard curve memorized in B·R·A·H·M·S KRYPTOR compact PLUS/KRYPTOR GOLD
Para utilizar con (equipo)B·R·A·H·M·S AFP KRYPTOR
IncluyeCalibrator kit, 6 vials
TipoCalibrador
Analitos detectablesalpha-fetoprotein
FormularioReady-to-Use (RTU)
Unit SizeBox with 6 vials
Contenido y almacenamiento
Calibrator kit, 6 vials, 2°C to 8°C