Dynabeads™ Protein G and Magnet Starter Pack
Invitrogen™
Dynabeads™ Protein G and Magnet Starter Pack
The Dynabeads Protein G and Magnet Starter Pack combines Dynabeads Protein G magnetic beads for immunoprecipitation with the magnet preferredRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSufficient For
10014D40 reactions
10017D100 reactions
Catalog number 10014D
Price (USD)
928.65
Online exclusive
1,020.00
Save 91.35
Each
Sufficient For:
40 reactions
Price (USD)
928.65
Online exclusive
1,020.00
Save 91.35
Each
The Dynabeads Protein G and Magnet Starter Pack combines Dynabeads Protein G magnetic beads for immunoprecipitation with the magnet preferred for use with 1.5 mL microcentrifuge tubes for purchasing convenience. Each mL of beads is sufficient for 20 reactions, and the magnet holds 16 x 1.5 mL microcentrifuge tubes. Dynabeads Protein G beads and the DynaMag-2 magnet are also available separately.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Ligand TypeProtein G
Quantity40 reactions
Shipping ConditionRoom Temperature
Sufficient For40 reactions
Product LineDynaMag, Dynabeads
TypeMagnet Starter Pack
Unit SizeEach
Contents & Storage
The product contains:
• 2 x 1 mL Dynabeads Protein G beads
• 1 x DynaMag-2 magnet
Store Dynabeads at 2–8°C.