The Dynabeads Protein G and Magnet Starter Pack combines Dynabeads Protein G magnetic beads for immunoprecipitation with the magnet preferredRead more
Have Questions?
Change view
Catalog Number
Sufficient For
10017D
100 reactions
10014D
40 reactions
2 Options
Catalog number 10017D
Price (USD)
1,226.65
Online exclusive
1,348.00
Save 121.35
Each
Sufficient For:
100 reactions
Price (USD)
1,226.65
Online exclusive
1,348.00
Save 121.35
Each
The Dynabeads Protein G and Magnet Starter Pack combines Dynabeads Protein G magnetic beads for immunoprecipitation with the magnet preferred for use with 1.5 mL microcentrifuge tubes for purchasing convenience. Each mL of beads is sufficient for 20 reactions, and the magnet holds 16 x 1.5 mL microcentrifuge tubes. Dynabeads Protein G beads and the DynaMag-2 magnet are also available separately.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Ligand TypeProtein G
Quantity100 reactions
Shipping ConditionRoom Temperature
Sufficient For100 reactions
Product LineDynaMag, Dynabeads
TypeMagnet Starter Pack
Unit SizeEach
Contents & Storage
The product contains: • 5 mL Dynabeads Protein G beads • 1 x DynaMag-2 magnet