Pacific Hemostasis™ Activated Partial Thromboplastin Time (APTT) Reagents
Actual product may vary
Thermo Scientific™

Pacific Hemostasis™ Activated Partial Thromboplastin Time (APTT) Reagents

Fisherbrand™ Pacific Hemostasis Activated Partial Thromboplastin Time (aPTT) Reagents are manufactured with up to two years extended shelf life.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionQuantity
100402TS
also known as 100402
APTT-XL (Ellagic Acid Activator)10 x 4 mL
100309CaCl2, 0.025M10 x 4 mL
100304TS
also known as 100304
CaCl2, 0.02M10 x 10 mL
100312TS
also known as 100312
KONTACT (Micronized Silica Activator)10 x 10 mL
100314TS
also known as 100314
CaCl2, 0.025M10 x 10 mL
100403aPTT-XL10 x 10 mL
100308KONTACT (Particulate Activator)10 x 4 mL
Catalog number 100402TS
also known as 100402
Price (USD)
-
Description:
APTT-XL (Ellagic Acid Activator)
Quantity:
10 x 4 mL
Identify acquired or congenital disorders to the intrinsic and common pathways with Thermo Scientific™ Pacific Hemostasis™ Activated Partial Thromboplastin Time (APTT) Reagents. Widely accepted as a means of monitoring patients on oral anticoagulant therapy such as Heparin, reagents are manufactured with up to two years extended shelf life and have excellent lot-to-lot consistency.
Features:
  • 10mL vials can complete ∽1000 full volume, single determinations; 4mL vials can complete ∽400 full volume, single determinations
  • Open vial stability: 30 days at 2° to 8°C

aPTT-XL (Ellagic Acid Activator) (Product No. 10-0402):

  • Superior factor sensitivity and responsiveness
  • Increased sensitivity to heparin and lupus-like inhibitors

KONTACT™ (Particulate Activator) (Product No. 01-0402):

  • Higher sensitivity to Factor VIII
  • Average Normal Values: 26 - 33 Seconds
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
DescriptionAPTT-XL (Ellagic Acid Activator)
Quantity10 x 4 mL
Shelf LifeUp to 2 Years
TypeActivated Partial Thromboplastin Time (APTT)
Unit SizeEach