MAX-iR FTIR Gas Analyzers
Thermo Scientific™
MAX-iR FTIR Gas Analyzers
The Thermo Scientific™ MAX-iR™ FTIR Gas Analyzer is designed to meet the challenges of in-line process, batch sampling, gas purity/certification, emissions testing, and ambient air monitoring applications.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoTipo de producto
101-0416Pureza del gas a baja presión (H2, Ar)
101-0343Prueba de la fuente
101-0344Pureza del gas a alta presión (N2)
101-0429Prueba de emisiones de turbinas de gas natural
Número de catálogo 101-0416
Precio (USD)
-
Tipo de producto:
Pureza del gas a baja presión (H2, Ar)
The Thermo Scientific™ MAX-iR™ FTIR Gas Analyzer is designed to meet the challenges of in-line process, batch sampling, gas purity/certification, emissions testing, and ambient air monitoring applications. Combined with Thermo Scientific™ StarBoost™ Enhanced Optical Technology, the MAX-iR analyzer enables users to achieve single-digit ppb detection limits for many applications. For ultra-high purity gas certification, the detection limits can be further reduced to mid-ppt.
MAX-iR FTIR Gas Analyzer Overview
The Thermo Scientific™ MAX-iR™ FTIR Gas Analyzer is designed to meet the challenges of in-line process, batch sampling, gas purity/certification, emissions testing, and ambient air monitoring applications. Combined with Thermo Scientific™ StarBoost™ Enhanced Optical Technology, the MAX-iR analyzer enables users to achieve single-digit ppb detection limits for many applications. For ultra-high purity gas certification, the detection limits can be further reduced to mid-ppt.
MAX-iR FTIR Gas Analyzer Features
  • High-performance analytical software specifically developed for real-time process and CEM environments.
  • Fully automated method control, DCS data output, alarm controls, remote access, and reporting tools.
  • Excellent signal-to-noise ratio without the use of liquid nitrogen.
  • Long-life vertical cavity surface-emitting laser (VCSEL) diode and a silicon carbide (SiC) IR source for increased longevity and dependability.
MAX-iR FTIR Gas Analyzer Applications
  • Inline process monitoring
  • Ambient air testing
  • Leak detection
  • Gas purity analysis
  • Continuous emissions monitoring (CEM)
  • Stationary source testing (RICE, turbines, cement kiln)
Especificaciones
Tipo de productoPureza del gas a baja presión (H2, Ar)
Tensión120 V
Unit SizeEach