Switch to Thermo Scientific DreamTaq DNA Polymerases for better results and added convenience.
<i>Taq</i> DNA Polymerase, recombinant
Invitrogen™

Taq DNA Polymerase, recombinant

Taq DNA Polymerase is a thermostable enzyme that synthesizes DNA from single-stranded templates in the presence of dNTPs and a primer.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. of Reactions
10342053100 Reactions
10342020500 Reactions
103420461500 Reactions
103421785000 Reactions
Catalog number 10342053
Price (USD)
74.65
Online exclusive
78.25
Save 3.60
Each
No. of Reactions:
100 Reactions
Request bulk or custom format
Price (USD)
74.65
Online exclusive
78.25
Save 3.60
Each
Taq DNA Polymerase, recombinant, is a thermostable enzyme that synthesizes DNA from single-stranded templates in the presence of dNTPs and a primer. This recombinant enzyme is purified from the cloned Thermus aquaticus DNA polymerase gene expressed in E. coli and consists of a single polypeptide with a molecular weight of 94 kDa. It has a 5' to 3' DNA polymerase activity and a 5' to 3' exonuclease activity.

Features

  • A choice of recombinant or native enzyme
  • Amplification of PCR products up to 5 kb in size
  • Enzyme is licensed and qualified for PCR

Applications

  • Amplification of DNA from complex genomic, viral, and plasmid templates
  • RT-PCR
  • Sequencing ssDNA
  • Cycle sequencing

Notes

  • One unit of Taq DNA Polymerase is the amount of enzyme required to incorporate 10 nmoles of deoxyribonucleotide into DNA in 30 minutes at 74°C.
  • For superior PCR performance, DreamTaq DNA Polymerase is recommended.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Exonuclease Activity5' - 3'
Fidelity (vs. Taq)1X
FormatStand-alone enzyme
Hot StartNo
No. of Reactions100 Reactions
Overhang3'-A
PolymeraseTaq DNA Polymerase
Product TypeDNA Polymerase
Quantity100 Units
Reaction FormatSeparate Components
Shipping ConditionDry Ice
Size (Final Product)5 kb or less
Starting MaterialDNA
Concentration5 U/μL
For Use With (Application)Standard PCR
GC-Rich PCR PerformanceLow
PCR MethodqPCR, Standard PCR
Reaction SpeedStandard
Unit SizeEach
Contents & Storage
• Taq DNA Polymerase (5 U/μL), 20 μL
• 10X PCR buffer, 1.5 mL
• 50 mM MgCl2, 1 mL

Store at -10°C to -30°C in a non-frost-free freezer.
Guaranteed stable for 6 months when stored properly.

Frequently asked questions (FAQs)

My oligonucleotide does not appear to be the right length when I checked by gel electrophoresis. Why is this?

Oligos should be run on a polyacrylamide gel containing 7 M urea and loaded with a 50% formamide solution to avoid compressions and secondary structures. Oligos of the same length and different compositions can electrophorese differently. dC's migrate fastest, followed by dA's, dT's, and then dG's. Oligos containing N's tend to run as a blurry band and generally have a problem with secondary structure.

The primers I am using worked for PCR initially, but over time, have stopped working. What happened?

Primers should be aliquoted for single use before PCR set-up. Heat just the aliquoted primers to 94 degrees for 1 min. Quick chill the primer on ice before adding to the PCR reaction. Some primers may anneal to themselves or curl up on themselves.

I don't see a pellet in my oligo tube order. Should I ask for a replacement?

The drying method dries the primer in a thin layer along the sidewalls of the tube instead of the bottom, therefore a pellet is not always visible and should still be ready to use.

There is a ball-shaped pellet at the bottom of my oligo tube. What is this and can I still use my oligo?

If the oligo was overheated, it will appear as a “ball”-shaped pellet attached to the bottom of the tube. This should not affect the quality of the oligo, and the oligo should be readily soluble in water.

There is a green color in my lyophilized oligo. Can I still use it?

If an oligo appears green in color, this is most likely due to ink falling into the tube. The oligo should still be fully functional. The color can be removed by doing an ethanol precipitation.

Citations & References (8)

Citations & References
Abstract
Both DNA and histone fold sequences contribute to archaeal nucleosome stability.
Authors: Bailey Kathryn A; Marc Frederic; Sandman Kathleen; Reeve John N;
Journal:J Biol Chem
PubMed ID:11751933
'The roles and interdependence of DNA sequence and archaeal histone fold structure in determining archaeal nucleosome stability and positioning have been determined and quantitated. The presence of four tandem copies of TTTAAAGCCG in the polylinker region of pLITMUS28 resulted in a DNA molecule with increased affinity (DeltaDeltaG of approximately 700 ... More
CCAAT/enhancer-binding proteins beta and delta mediate the repression of gene transcription of cartilage-derived retinoic acid-sensitive protein induced by interleukin-1 beta.
Authors: Okazaki Ken; Li Jian; Yu Hua; Fukui Naoshi; Sandell Linda J;
Journal:J Biol Chem
PubMed ID:12072435
'Cartilage-derived retinoic acid-sensitive protein (CD-RAP) is a secreted protein expressed by chondrocytes; the expression is repressed by interleukin 1 beta (IL-1 beta). To investigate the transcriptional mechanism, by which CD-RAP expression is suppressed by IL-1 beta, deletion constructs of the mouse CD-RAP promoter were transfected into rat chondrocytes treated with ... More
alpha-Methylacyl coenzyme A racemase as a tissue biomarker for prostate cancer.
Authors: Rubin Mark A; Zhou Ming; Dhanasekaran Saravana M; Varambally Sooryanarayana; Barrette Terrence R; Sanda Martin G; Pienta Kenneth J; Ghosh Debashis; Chinnaiyan Arul M;
Journal:JAMA
PubMed ID:11926890
'Edited due to space constraints: CONTEXT: Molecular profiling of prostate cancer has led to the identification of candidate biomarkers and regulatory genes. Discoveries from these genome-scale approaches may have applicability in the analysis of diagnostic prostate specimens. OBJECTIVES: To determine the expression and clinical utility of alpha-methylacyl coenzyme A racemase ... More
The Salmonella enterica serovar typhimurium-encoded type III secretion systems can translocate Chlamydia trachomatis proteins into the cytosol of host cells.
Authors:Ho TD, Starnbach MN,
Journal:Infect Immun
PubMed ID:15664932
Chlamydia trachomatis is an obligate, intracellular pathogen that is a major cause of preventable blindness and infertility worldwide. Although the published genome sequence suggests that C. trachomatis encodes a type III secretion system, the lack of genetic tools for studying Chlamydia has hindered the examination of this potentially important class ... More
Requirement for either a host- or pectin-induced pectate lyase for infection of pisum sativum by nectriahematococca.
Authors:Rogers LM, Kim YK, Guo W, Gonzalez-Candelas L, Li D, Kolattukudy PE
Journal:Proc Natl Acad Sci U S A
PubMed ID:10931947
Fungal pathogens usually have multiple genes that encode extracellular hydrolytic enzymes that may degrade the physical barriers in their hosts during the invasion process. Nectria hematococca, a plant pathogen, has two inducible pectate lyase (PL) genes (pel) encoding PL that can help degrade the carbohydrate barrier in the host. pelA ... More